Learn More
ITGA3 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA547398
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant mouse (rm) Integrin alpha 2, rmIntegrin alpha 4, rmIntegrin alpha 5, rmIntegrin alpha E, rmIntegrin alpha M, rmIntegrin alpha V and rmIntegrin alpha X is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
ITGA3 Polyclonal antibody specifically detects ITGA3 in Mouse samples. It is validated for Flow Cytometry, Western Blot.Spezifikation
ITGA3 | |
Polyclonal | |
Unconjugated | |
ITGA3 | |
AA407068; alpha 3 subunit of VLA-3 receptor; Alpha3 integrin; antigen CD49C; antigen identified by monoclonal antibody J143; CD49 antigen-like family member C; CD49c; CD49c antigen; FLJ34631; FLJ34704; FRP-2; Galactoprotein B3; GAPB3; GAP-B3; ILNEB; integrin alpha 3; Integrin alpha3; Integrin alpha-3; Integrin alpha-3 heavy chain; Integrin alpha-3 light chain; integrin subunit alpha 3; integrin, alpha 3; integrin, alpha 3 (antigen CD49C, alpha 3 subunit of VLA-3 receptor); ITGA3; MSK18; testicular tissue protein Li 96; testicular tissue protein Li 98; VCA-2; very late activation protein 3 receptor, alpha-3 subunit; VL3A; VLA-3 subunit alpha; VLA3a | |
Goat | |
Antigen affinity chromatography | |
RUO | |
16400 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q62470 | |
ITGA3 | |
Sf 21-derived recombinant mouse Integrin alpha 3/CD49c Phe33-Glu993 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |