missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITGA3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA532881
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey, Bovine (100%) Marmoset, Mouse, Dog, Bat, Hamster, Elephant, Horse, Opossum (94%) Rabbit (89%).
ITGA3 Polyclonal antibody specifically detects ITGA3 in Bovine, Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
ITGA3 | |
Polyclonal | |
Unconjugated | |
ITGA3 | |
AA407068; alpha 3 subunit of VLA-3 receptor; Alpha3 integrin; antigen CD49C; antigen identified by monoclonal antibody J143; CD49 antigen-like family member C; CD49c; CD49c antigen; FLJ34631; FLJ34704; FRP-2; Galactoprotein B3; GAPB3; GAP-B3; ILNEB; integrin alpha 3; Integrin alpha3; Integrin alpha-3; Integrin alpha-3 heavy chain; Integrin alpha-3 light chain; integrin subunit alpha 3; integrin, alpha 3; integrin, alpha 3 (antigen CD49C, alpha 3 subunit of VLA-3 receptor); ITGA3; MSK18; testicular tissue protein Li 96; testicular tissue protein Li 98; VCA-2; very late activation protein 3 receptor, alpha-3 subunit; VL3A; VLA-3 subunit alpha; VLA3a | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3675, 508490 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
F1MMS9, P26006 | |
ITGA3 | |
Synthetic 18 amino acid peptide from N-terminus of human ITGA3 / CD49c. | |
50 μg | |
Primary | |
Bovine, Human | |
Antibody | |
IgG |