Learn More
ITGA8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA542739
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: GEKQTEVAPA SYDDSYLGYS VAAGEFTGDS QQELVAGIPR GAQNFGYVSI Sequence homology: Cow: 93%; Dog: 86%; Guinea Pig: 91%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 79%
ITGA8 Polyclonal antibody specifically detects ITGA8 in Human samples. It is validated for Western Blot.Spezifikation
ITGA8 | |
Polyclonal | |
Unconjugated | |
ITGA8 | |
AI447669; integrin alpha 8; integrin alpha-8; Integrin alpha-8 heavy chain; Integrin alpha-8 light chain; integrin subunit alpha 8; integrin, alpha 8; ITGA8; RGD1564327; zgc:172265 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
8516 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P53708 | |
ITGA8 | |
Synthetic peptide directed towards the N-terminal of human ITGA8 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |