missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11a Monoclonal Antibody (MEM-25), APC, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen A15699
Weitere Details : Netto-Gewicht : 0.00280kg
Beschreibung
CD11a Monoclonal antibody specifically detects CD11a in Human samples. It is validated for Flow Cytometry.Spezifikation
CD11a, itgal | |
Monoclonal | |
APC | |
ITGAL | |
(p180); alpha L integrin; antigen CD11A (p180); antigen CD11A (p180), lymphocyte function-associated antigen 1, alpha polypeptide; antigen CD11A p180; CD11 antigen-like family member A; CD11a; integrin alpha L; integrin alphaL; integrin alpha-L; integrin alpha-L precursor; integrin gene promoter; integrin subunit alpha L; integrin surface receptor; integrin, alpha L; integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen 1; alpha polypeptide); ITGAL; Leukocyte adhesion glycoprotein LFA-1 alpha chain; leukocyte function-associated molecule 1 alpha chain; LFA-1; LFA-1 alpha; LFA1A; LFA-1A; Ly-15; Ly-21; lymphocyte antigen 15; lymphocyte function associated antigen 1, alpha polypeptide; lymphocyte function-associated antigen 1; lymphocyte function-associated antigen 1, alpha polypeptide; lymphocyte function-associated antigen-1 | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
3683 | |
4° C | |
Liquid |
Flow Cytometry | |
MEM-25 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P20701 | |
ITGAL | |
Leukocytes from a patient suffering from a LGL-type leukaemia | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |