missing translation for 'onlineSavingsMsg'
Learn More

anti-ITGAM/CD11b APC, Clone: ICRF44, Invitrogen™

ITGAM/CD11b Monoclonal antibody A18613 from Invitrogen specifically detects ITGAM/CD11b in Human samples. It is validated for Flow Cytometry.

Marke:  Invitrogen A18613

Weitere Details : Netto-Gewicht : 0.00280kg

Artikelnummer. 15266174

Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen
Alternative Produkte anzeigen »


Beschreibung

Beschreibung

ITGAM/CD11b Monoclonal antibody A18613 from Invitrogen specifically detects ITGAM/CD11b in Human samples. It is validated for Flow Cytometry.

CD11b is implicated in various adhesive interactions of monocytes, macrophages and granulocytes as well as in mediating the uptake of complement coated particles. It is identical to CR3, the receptor for the iC3b fragment of the third complement component. It probably recognizes the RGD peptide in C3b. CD11b is also a receptor for fibrinogen, factor X, and ICAM1. It recognizes P1 and P2 peptides of fibrinogen gamma chain. The Mac1 CD11b antigen is present on macrophages, granulocytes, natural killer cells, and blood monocytes. CD11b is expressed on 8% of spleen cells, 44% of bone marrow cells, and less than 1% of thymocytes, and is commonly used as a microglial marker in nervous tissue.
Host Species: Mouse
Cross Reactivity: Human
Isotype: IgG1, κ
Clone: ICRF44
Immunogen: Rheumatoid synovial cells and human monocytes.
Formulation: 25 Tests Purified Antibody (0.2mg/ml) in PBS, with 0.1% gelatin and 0.09% sodium azide; pH7.2
Synonyms: CD11B, CR3A, MAC-1, MAC1A,mgC117044, MO1A, SLEB6, Integrin alpha M
Uniprot Accession: P11215
Entrez GeneID: 3684
Gene Symbol: ITGAM
This product detects ITGAM/CD11b in Human samples. This product has been successfully used in Flow Cytometry procedures. The immunogen is Rheumatoid synovial cells and human monocytes.
Spezifikation

Spezifikation

ITGAM/CD11b
Monoclonal
APC
P11215
ITGAM
Rheumatoid synovial cells and human monocytes.
Primary
Human
Purified
Flow Cytometry
ICRF44
25 Tests Purified Antibody (0.2mg/ml) in PBS, with 0.1% gelatin and 0.09% sodium azide; pH7.2
CD11B, CR3A, MAC-1, MAC1A,mgC117044, MO1A, SLEB6, Integrin alpha M
Mouse
25 Tests
3684
Store at 4° C
IgG1 κ
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote