Learn More
ITGB1 Monoclonal Antibody (3B6), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA517103
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-17103 targets ITGB1 in FACS, IHC, indirect ELISA, and WB applications and shows reactivity with Human and Non-human primate samples. The MA5-17103 immunogen is purified recombinant fragment of human ITGB1 expressed in E. Coli. MA5-17103 detects ITGB1 which has a predicted molecular weight of approximately 88.4kDa.
ITGB1 Monoclonal antibody specifically detects ITGB1 in Human, Mouse, Non-human Primate samples. It is validated for ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
ITGB1 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
P05556, P09055 | |
ITGB1 | |
Purified recombinant fragment of human ITGB1 expressed in E. Coli. | |
100 μg | |
Primary | |
Human, Mouse, Non-human Primate | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
3B6 | |
Unconjugated | |
ITGB1 | |
4633401G24Rik; AA409975; AA960159; Beta OL; beta oligodendroglia; CD29; CSAT antigen; ENSMUSG00000051907; Fibronectin receptor subunit beta; FNRB; Glycoprotein IIa; Gm9863; gpIIa; integrin beta 1; integrin beta 1 (fibronectin receptor beta); integrin beta 1A precursor; Integrin beta1; integrin beta-1; integrin beta-1 subunit; integrin beta1D; integrin precursor; integrin subunit beta 1; integrin VLA-4 beta subunit; integrin VLA-4 subunit beta; integrin, beta 1; integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includes MDF2, MSK12); ITBG1D; ITGB1; JG22 antigen; MDF2; MSK12; RGD-receptor; very late activation protein, beta polypeptide; VLA-4 subunit beta; VLAB; VLA-BETA | |
Mouse | |
Protein G | |
RUO | |
16412, 3688 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |