Learn More
ITGB1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA532888
Weitere Details : Netto-Gewicht : 0.01000kg
Benachrichtigungen:
g
Beschreibung
Percent identity with other species by BLAST analysis: Human, Gorilla, Orangutan, Gibbon, Monkey, Marmoset, Sheep, Dog, Bovine, Cat, Elephant, Panda, Horse, Pig (100%); Platypus (93%); Bat, Opossum (86%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
ITGB1 Polyclonal antibody specifically detects ITGB1 in Canine, Equine, Human, Non-human Primate, Ovine, Porcine samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
ITGB1 | |
Polyclonal | |
Unconjugated | |
ITGB1 | |
4633401G24Rik; AA409975; AA960159; Beta OL; beta oligodendroglia; CD29; CSAT antigen; ENSMUSG00000051907; Fibronectin receptor subunit beta; FNRB; Glycoprotein IIa; Gm9863; gpIIa; integrin beta 1; integrin beta 1 (fibronectin receptor beta); integrin beta 1A precursor; Integrin beta1; integrin beta-1; integrin beta-1 subunit; integrin beta1D; integrin precursor; integrin subunit beta 1; integrin VLA-4 beta subunit; integrin VLA-4 subunit beta; integrin, beta 1; integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includes MDF2, MSK12); ITBG1D; ITGB1; JG22 antigen; MDF2; MSK12; RGD-receptor; very late activation protein, beta polypeptide; VLA-4 subunit beta; VLAB; VLA-BETA | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100055024, 3688, 397019, 443141, 477956 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 0.1% sodium azide | |
B0FYY4, P05556, Q9GLP0 | |
ITGB1 | |
Synthetic 14 amino acid peptide from extracellular domain of human Integrin Beta 1. | |
50 μg | |
Primary | |
Canine, Equine, Human, Non-human Primate, Ovine, Porcine | |
Antibody | |
IgG |