Learn More
CD61 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA532889
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey (100%); Marmoset (93%); Dog, Panda (86%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
CD61 Polyclonal antibody specifically detects CD61 in Equine, Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
CD61 | |
Polyclonal | |
Unconjugated | |
ITGB3 | |
antigen CD61; BDPLT16; BDPLT2; beta3 integrin; CD61; EGK_08849; glycoprotein GPIIIa; glycoprotein IIIa; GP3A; GPIIIa; GT; INGRB3; integrin beta 3; integrin beta 3 (Cd61); integrin beta-3; integrin beta-3 subunit; integrin subunit beta 3; integrin, beta 3; integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61); ITGB3; platelet glycoprotein IIIa; platelet glycoprotein IIIa integrin subunit beta 3; platelet gpIIIa; platelet membrane glycoprotein IIIa | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100009709, 3690 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.9 mg/mL | |
PBS with 0.1% sodium azide | |
P05106 | |
ITGB3 | |
Synthetic 14 amino acid peptide from internal region of human CD61. | |
50 μg | |
Primary | |
Equine, Human | |
Antibody | |
IgG |