missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITK Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA514782
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
ITK Polyclonal antibody specifically detects ITK in Human samples. It is validated for Western Blot.Spezifikation
ITK | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q08881 | |
ITK | |
KLH conjugated synthetic peptide between 169-198 amino acids from the central region of human ITK | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
ITK | |
EMT; homolog of mouse T-cell itk/tsk; IL2 inducible T cell kinase; IL2 inducible T-cell kinase; IL-2-inducible T cell kinase; IL-2-inducible T-cell kinase; IL2-inducible T-cell kinase; interleukin-2-inducible T cell kinase; interleukin-2-inducible T-cell kinase; ITK; Kinase EMT; kinase TLK; LPFS1; LYK; MGC126257; MGC126258; PSCTK2; T-cell-specific kinase; Tcsk; Tlk; Tsk; Tyrosine-protein kinase ITK/TSK; tyrosine-protein kinase LYK | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
3702 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |