missing translation for 'onlineSavingsMsg'
Learn More

IVNS1ABP Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA531277

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 13405797

  • 473.00 € / 100 Mikroliter
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-7. Predicted reactivity: Mouse (97%), Rat (97%), Pig (95%), Rhesus Monkey (100%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

IVNS1ABP Polyclonal antibody specifically detects IVNS1ABP in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.
Spezifikation

Spezifikation

IVNS1ABP
Polyclonal
Unconjugated
IVNS1ABP
1190004M08Rik; 1700126I16Rik; AA960440; ARA3; aryl hydrocarbon receptor-associated 3; aryl hydrocarbon receptor-associated protein 3; FLARA3; HSPC068; influenza virus NS1A binding protein; influenza virus NS1A-binding protein; Influenza virus NS1A-binding protein homolog; Ivns1abp; kelch family protein Nd1; kelch family protein Nd1-L; kelch-like family member 39; Kelch-like protein 39; KIAA0850; KLHL39; mKIAA0850; NCX downstream gene 1; ND1; Nd1L; Nd1-L; ND1-L2; Nd1S; Nd1-S; NS1; NS-1; NS1-binding protein; NS1-binding protein homolog; Ns1bp; NS1-BP
Rabbit
Antigen affinity chromatography
RUO
10625, 117198
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.57 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Q920Q8, Q9Y6Y0
IVNS1ABP
Recombinant fragment corresponding to a region within amino acids 415 and 640 of Human IVNS1ABP
100 μL
Primary
Human, Mouse
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote