missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kaiso Monoclonal Antibody (6F), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen 358500
Weitere Details : Netto-Gewicht : 0.50000kg
Beschreibung
Kaiso Monoclonal antibody specifically detects Kaiso in Canine, Chicken, Human, Mouse, Rabbit, Rat samples. It is validated for ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot.Spezifikation
Kaiso | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
Q86T24, Q8BN78 | |
ZBTB33 | |
Fusion protein encompassing the N-terminus of mouse Kaiso | |
100 μg | |
Primary | |
Canine, Chicken, Human, Mouse, Rabbit, Rat | |
Antibody | |
IgG1 |
ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot | |
6F | |
Unconjugated | |
ZBTB33 | |
AW260253; E130014G12Rik; Kaiso; kaiso transcription factor; RGD1566309; RP23-322E15.9; Transcriptional regulator Kaiso; WUGSC:H_DJ525N14.1; Zbtb33; zinc finger and BTB domain containing 33; zinc finger and BTB domain-containing protein 33; zinc finger transcription factor Kaiso; ZNF348; ZNF-kaiso | |
Mouse | |
Protein A | |
RUO | |
10009, 100348939, 100685062, 422367, 501506, 56805 | |
-20°C | |
Liquid |