missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kalirin Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518214
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Kalirin Polyclonal antibody specifically detects Kalirin in Human samples. It is validated for Western Blot.Spezifikation
Kalirin | |
Polyclonal | |
Unconjugated | |
KALRN | |
2210407G14Rik; ARHGEF24; AV235988; CHD5; CHDS5; cyclin B1; DUET; DUO; E530005C20Rik; FLJ12332; FLJ16443; FLJ18196; FLJ18623; Gm539; HAPIP; hCG_2022551; huntingtin-associated protein interacting protein (duo); huntingtin-associated protein-interacting protein; Kalirin; kalirin, RhoGEF kinase; kalrn; PAM COOH-terminal interactor protein 10; P-CIP10; Protein Duo; RhoGEF kinase; serine/threonine kinase with Dbl- and pleckstrin homology domain; serine/threonine kinase with Dbl- and pleckstrin homology domains; serine/threonine-protein kinase with Dbl- and pleckstrin homology domain; TRAD | |
Goat | |
Antigen affinity chromatography | |
RUO | |
8997 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O60229 | |
KALRN | |
Synthetic peptide sequence (RWHLGPGDPFSTYV) corresponding to the C-terminus amino acids of KALRN | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |