missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kallikrein 11 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA146127
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Kallikrein 11 Polyclonal antibody specifically detects Kallikrein 11 in Human samples. It is validated for Western Blot.Spezifikation
Kallikrein 11 | |
Polyclonal | |
Unconjugated | |
Klk11 | |
2310015I08Rik; Hippostasin; hK11; kallikrein 11; kallikrein related peptidase 11; kallikrein related-peptidase 11; Kallikrein11; kallikrein-11; Kallikrein-11 inactive chain 1; Kallikrein-11 inactive chain 2; kallikrein-related peptidase 11; KLK11; protease, serine, 20; Prss20; Serine protease 20; TLSP; Trypsin-like protease; UNQ649/PRO1279 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11012 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q9UBX7 | |
Klk11 | |
Residues 68-82 of the 35 kDa human hippostasin/Kallikrein 11protein. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |