Learn More
Kallikrein 15 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA14211
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-4211 detects the Kallikrein-15 protein in human samples. PA1-4211 has been successfully used in Western blot procedures. The PA1-4211 immunogen is a synthetic peptide corresponding to residues H(75) N L R K R D G P E Q L R(87) of human Kallikrein-15.
Kallikrein 15 Polyclonal antibody specifically detects Kallikrein 15 in Human samples. It is validated for Western Blot.Spezifikation
Kallikrein 15 | |
Polyclonal | |
Unconjugated | |
KLK15 | |
ACO; ACO protease; HSRNASPH; kallikrein 15; kallikrein related peptidase 15; kallikrein related-peptidase 15; kallikrein-15; kallikrein-like serine protease; kallikrein-related peptidase 15; KLK15; KLK-15; prostinogen | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55554 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q9H2R5 | |
KLK15 | |
Synthetic peptide corresponding to residues H(75) N L R K R D G P E Q L R(87) of human Kallikrein-15. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |