missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kallikrein 5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA121722
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-21722 detects Kallikrein-L2 in Human samples.
Kallikrein 5 Polyclonal antibody specifically detects Kallikrein 5 in Human samples. It is validated for Western Blot.Spezifikation
Kallikrein 5 | |
Polyclonal | |
Unconjugated | |
Klk5 | |
1110030O19Rik; HK5; kallikrein 5; Kallikrein L2; Kallikrein like 2; kallikrein related peptidase 5; kallikrein related-peptidase 5; Kallikrein5; kallikrein-5; kallikrein-5-like; kallikrein-like protein 2; kallikrein-related peptidase 5; KLK 5; KLK L2; KLK5; KLK-5; KLKL 2; KLKL2; KLK-L2; LOC102546758; SCTE; Stratum corneum tryptic enzyme; UNQ570/PRO1132 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25818 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q9Y337 | |
Klk5 | |
Synthetic peptide: KFTKWIQETIQANS, corresponding to amino acids 280-293 of Human KLKL2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |