Learn More
TRIM28 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA1852
Additional Details : Netto-Gewicht : 0.01000kg
Description
PA1-852 detects KAP-1 (Kruppel-associated box-associated protein 1) in human, mouse and rat samples. PA1-852 has been successfully used in Western blot, ICC/IF and IHC (P) procedures. By Western blot, this antibody detects an ∽87 kDa protein representing recombinant human KAP-1. This antibody also detects a smaller protein that is believed to be KAP-1 degradation product. In immunofluorescent applications, this antibody primarily reacts with mouse or rat and shows non-specific staining in HeLa cells. The PA1-852 antigen is a recombinant human KAP-1.
TRIM28 Polyclonal antibody specifically detects TRIM28 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
TRIM28 | |
Polyclonal | |
Unconjugated | |
TRIM28 | |
AA408787; E3 SUMO-protein ligase TRIM28; KAP1; KAP-1; KRAB [Kruppel-associated box domain]-associated protein 1; KRAB-A-interacting protein; KRAB-associated protein 1; KRAB-interacting p; KRAB-interacting protein 1; Krip1; KRIP-1; MommeD9; Nuclear corepressor KAP-1; PPP1R157; protein phosphatase 1, regulatory subunit 157; RING finger protein 96; RING-type E3 ubiquitin transferase TIF1-beta; RNF96; TF1B; tif1 beta; Tif1b; Tif1beta; TIF1-beta; Transcription intermediary factor 1-beta; transcriptional intermediary factor 1, beta; transcriptional intermediary factor 1-beta; Trim28; tripartite motif; tripartite motif containing 28; tripartite motif protein 28; tripartite motif-containing 28; tripartite motif-containing protein 28 | |
Rabbit | |
Ammonium sulfate precipitation | |
RUO | |
10155, 116698, 21849 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Conc. Not Determined | |
PBS with 0.05% sodium azide | |
O08629, Q13263, Q62318 | |
TRIM28 | |
Recombinant human KAP-1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |