Learn More
OPRK1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA124535
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: Rat brain. Predicted reactivity: Peptide sequence identity/similarity to Mouse: 88/100%. Rat: 88/100%.. PA1-24535 has been successfully used in Immunohistochemistry (paraffin) procedures.
OPRK1 Polyclonal antibody specifically detects OPRK1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
OPRK1 | |
Polyclonal | |
Unconjugated | |
OPRK1 | |
kappa opioid receptor; kappa-type opioid receptor; KOPR1; KOR; K-OR-1; KOR-1; MSL-1; Opiate receptor, kappa-1; opioid receptor kappa 1; opioid receptor, kappa 1; opioid receptor, kappa 2; OPRK; OPRK1; Oprk2; Ork1; R21; Ror-d | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18387, 29335, 4986 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P33534, P34975, P41145 | |
OPRK1 | |
Synthetic 18 amino acid peptide from C-terminus of human Kappa Opioid Receptor. | |
25 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |