missing translation for 'onlineSavingsMsg'
Learn More

Kappa Light Chain Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA516647

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11529431

  • 414.00 € / 500 Mikroliter
Verfügbar ab: 15-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

PA5-16647 targets Kappa Light Chain in IHC (P) applications and shows reactivity with Human, baboon, monkey and rat samples. Does not react with dog samples. Staining of formalin-fixed tissues requires boiling tissue sections in 10mM citrate buffer, pH 6.0, for 10-20 minutes followed by cooling at room temperature for 20 minutes. The PA5-16647 immunogen is kappa light chains purified from human Bence Jones proteins.

Kappa Light Chain Polyclonal antibody specifically detects Kappa Light Chain in Human samples. It is validated for Immunohistochemistry (Paraffin).
Spezifikation

Spezifikation

Kappa Light Chain
Polyclonal
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
P01602, P01834, P0DOX7
IGK, IGKC, IGKV1-5
kappa light chains purified from human Bence Jones proteins
500 μL
Primary
Human
Antibody
IgG
Immunohistochemistry (Paraffin)
Unconjugated
IGK
D4Mit8; HCAK1; ig kappa chain c region; Igk; Igk@; Igkc; Igk-C; IGKCD; IGKV; IGKV15; IGKV1-5; Immunoglobulin K; immunoglobulin kappa chain complex; immunoglobulin kappa constant; immunoglobulin kappa light chain; immunoglobulin kappa locus; immunoglobulin kappa variable 1-5; kappa; Kappa LC; Km; L12; L12a; LOC500183; similar to NGF-binding Ig light chain; V1
Rabbit
purified
RUO
28299, 3514, 50802
4° C
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote