Learn More
Mouse anti-Human Kappa Light Chain Secondary Antibody, Invitrogen™
Mouse Monoclonal Secondary Antibody
Marke: Invitrogen MA110385
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody will not cross-react with hamster, goat, guinea pig, rabbit or sheep.
Human Kappa Light Chain Monoclonal antibody specifically detects Human Kappa Light Chain in Human samples. It is validated for ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).Spezifikation
Human Kappa Light Chain | |
Monoclonal | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
not available | |
100 μg | |
Secondary | |
4° C, do not freeze | |
IgG1 |
ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
A8B5 | |
Unconjugated | |
Mouse | |
Protein A | |
RUO | |
Human | |
Liquid |