missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIP60 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA511216
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with mouse and rat based on sequence homology.
TIP60 Polyclonal antibody specifically detects TIP60 in Human, Mouse samples. It is validated for Western Blot.Spezifikation
TIP60 | |
Polyclonal | |
Unconjugated | |
KAT5 | |
60 kDa Tat-interactive protein; AI839539; cPLA(2)-interacting protein; CPLA2; cPLA2 interacting protein; ESA1; histone acetyltransferase HTATIP; Histone acetyltransferase KAT5; HIV-1 Tat interactive protein; HIV-1 tat interactive protein 1, 60 kDa homolog; HIV-1 tat interactive protein 1, homolog; HIV-1 Tat interactive protein 60 kD; HIV-1 Tat interactive protein, 60 kD; HIV-1 Tat interactive protein, 60kDa; HIV-1 tat interactive protein, homolog; HTATIP; Htatip1; K(lysine) acetyltransferase 5; K-acetyltransferase 5; Kat5; Lysine acetyltransferase 5; PLIP; Tat interacting protein, 60kDa; Tat-interactive protein-60; TIP; Tip55; Tip60; TIP60(HTATIP); Tip60b; ZC2HC5 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
10524, 81601 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide | |
Q8CHK4, Q92993 | |
KAT5 | |
KLH conjugated synthetic peptide between 33-64 amino acids from the N-terminal region of human KAT5 / Tip60/HTATIP | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |