missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KBTBD11 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA546490
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: GGPTGLQPFR CAALDGAIYC VSRAGTWRFQ PAREGEAGGD AGQGGGFEAL Sequence homology: Human: 100%
KBTBD11 Polyclonal antibody specifically detects KBTBD11 in Human samples. It is validated for Western Blot.Spezifikation
KBTBD11 | |
Polyclonal | |
Unconjugated | |
KBTBD11 | |
2900016B01Rik; 4930465M17Rik; Chronic myelogenous leukemia-associated protein; CMLAP; Kbtbd11; Kelch domain-containing protein 7B; kelch repeat and BTB (POZ) domain containing 11; kelch repeat and BTB domain containing 11; kelch repeat and BTB domain-containing protein 11; KIAA0711; KLHDC7C; mKIAA0711 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
9920 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O94819 | |
KBTBD11 | |
Synthetic peptide directed towards the C-terminal of human KBTBD11 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |