missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KChIP3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA511665
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
KChIP3 Polyclonal antibody specifically detects KChIP3 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
KChIP3 | |
Polyclonal | |
Unconjugated | |
Kcnip3 | |
4933407H12Rik; AI413860; A-type potassium channel modulating protein 3; A-type potassium channel modulatory protein 3; Calsenilin; calsenilin presenilin-binding protein EF hand transcription factor; calsenilin, presenilin binding protein, EF hand transcription factor; calsenilin, presenilin-binding protein, EF hand transcription fa; calsenilin, presenilin-binding protein, EF hand transcription factor; CSEN; downstream regulatory element-antagonist modulator; DREAM; DRE-antagonist modulator; KChIP3; KCNIP3; Kv channel interacting protein 3; Kv channel interacting protein 3, calsenilin; kv channel-interacting protein 3; potassium channel interacting protein 3; potassium voltage-gated channel interacting protein 3; R74849; rKChIP3 | |
Rabbit | |
Protein G | |
RUO | |
30818, 56461 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1.6 mg/mL | |
PBS with 0.09% sodium azide | |
Q9QXT8, Q9Y2W7 | |
Kcnip3 | |
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human KChIP3 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |