Learn More
KVbeta1 (KCNAB1) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA541019
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: VPESSRASLK CYQWLKERIV SEEGRKQQNK LKDLSPIAER LGCTLPQLAV Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 77%
KVbeta1 (KCNAB1) Polyclonal antibody specifically detects KVbeta1 (KCNAB1) in Human, Mouse samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
KVbeta1 (KCNAB1) | |
Polyclonal | |
Unconjugated | |
KCNAB1 | |
AKR6A3; Akr8a8; hKvb3; hKvBeta3; K(+) channel subunit beta-1; K+ channel Beta1a chain; KCNA1B; KCNAB1; Kvb1; Kvb1.3; KVBETA1; Kv-beta-1; Kvbeta1.1; KvBeta3; mKv(beta)1; potassium channel beta 3 chain; potassium channel beta3 subunit; potassium channel shaker chain beta 1a; potassium channel subunit Kv beta 1; potassium channel subunit Kv beta 1.1; potassium channel subunit Kv beta 1.2; potassium channel subunit Kv beta 1.3; potassium channel, voltage gated subfamily A regulatory beta subunit 1; potassium channel, voltage-gated shaker related subfamily A regulatory beta subunit 1; potassium voltage gated channel shaker related subfamily beta member 1; potassium voltage gated channel, shaker related subfamily, beta member 1; potassium voltage-gated channel shaker-related subfamily beta member 1; potassium voltage-gated channel subfamily A member regulatory beta subunit 1; potassium voltage-gated channel, shaker-related subfamily, beta member 1; voltage-gated potassium channel beta subuni | |
Rabbit | |
Affinity Chromatography | |
RUO | |
16497, 7881 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P63143, Q14722 | |
KCNAB1 | |
Synthetic peptide directed towards the C-terminal of human KCNAB1 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |