missing translation for 'onlineSavingsMsg'
Learn More

KV3.1 (KCNC1) Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA542894

Weitere Details : Netto-Gewicht : 0.00010kg

Artikelnummer. 15575762

  • 473.00 € / 100 Mikroliter
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Peptide sequence: TYRSTLRTLP GTRLAWLAEP DAHSHFDYDP RADEFFFDRH PGVFAHILNY Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%

KV3.1 (KCNC1) Polyclonal antibody specifically detects KV3.1 (KCNC1) in Human samples. It is validated for Western Blot.
Spezifikation

Spezifikation

KV3.1 (KCNC1)
Polyclonal
Unconjugated
KCNC1
C230009H10Rik; EPM7; FLJ41162; FLJ42249; FLJ43491; KCNC1; Kcr2-1; Krc2 1; KShIIIB; KV3.1; KV4; MGC129855; NGK2; NGK2-KV4; Potassium channel gene 1 (alternative splicing product described in Luneau et al 1991); potassium channel, voltage gated Shaw related subfamily C, member 1; potassium channel, voltage gated Shaw-related subfamily C, member 1; potassium voltage gated channel, Shaw-related subfamily, member 1; potassium voltage-gated channel subfamily C member 1; potassium voltage-gated channel, Shaw-related subfamily, member 1; RAW2; Shaw; Shaw-related voltage-gated potassium channel protein 1; voltage-gated potassium channel KCNC1; voltage-gated potassium channel protein KV3.1; voltage-gated potassium channel subunit Kv3.1; Voltage-gated potassium channel subunit Kv4
Rabbit
Protein A
RUO
3746
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
1 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
P48547
KCNC1
Synthetic peptide directed towards the N-terminal of human KCNC1
100 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote