missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kir5.1 (KCNJ16) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA541051
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: RESCTSDTKA RRRSFSAVAI VSSCENPEET TTSATHEYRE TPYQKALLTL Sequence homology: Dog: 86%; Horse: 93%; Human: 100%; Pig: 79%; Rabbit: 86%
Kir5.1 (KCNJ16) Polyclonal antibody specifically detects Kir5.1 (KCNJ16) in Human samples. It is validated for Western Blot.Spezifikation
Kir5.1 (KCNJ16) | |
Polyclonal | |
Unconjugated | |
Kcnj16 | |
6430410F18Rik; AI132396; BIR9; Inward rectifier K(+) channel Kir5.1; inward rectifier K+ channel KIR5.1; inward rectifier potassium channel 16; inwardly-rectifying potassium channel Kir5.1; Kcnj16; KIR5.1; MGC33717; potassium channel, inwardly rectifying subfamily J member 16; potassium channel, inwardly rectifying subfamily J, member 16; potassium inwardly rectifying channel subfamily J member 16; potassium inwardly-rectifying channel J16; potassium inwardly-rectifying channel, subfamily J, member 16; potassium voltage-gated channel subfamily J member 16 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
3773 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9NPI9 | |
Kcnj16 | |
Synthetic peptide directed towards the middle region of human KCNJ16 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |