missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kir3.2 (KCNJ6) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA533869
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Kir3.2 (KCNJ6) Polyclonal antibody specifically detects Kir3.2 (KCNJ6) in Canine, Equine, Hamster, Human, Mouse, Non-human Primate, Porcine, Rat samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
Kir3.2 (KCNJ6) | |
Polyclonal | |
Unconjugated | |
KCNJ6 | |
BIR1; G protein-activated inward rectifier potassium channel 2; Girk2; GIRK-2; hiGIRK2; Inward rectifier K(+) channel Kir3.2; inward rectifier potassium channel KIR3.2; KATP2; KATP-2; KCNJ6; Kcnj7; KIR3.2; KPLBS; MGC126596; Potassium channel, inwardly rectifying subfamily J member 6; potassium channel, inwardly rectifying subfamily J, member 6; potassium inwardly-rectifying channel J6; potassium inwardly-rectifying channel, subfamily J, member 6; potassium voltage-gated channel subfamily J member 6; W; weaver; wv | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100060665, 100515928, 100763599, 16522, 25743, 3763, 487756 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P48051, P48542, P48550 | |
KCNJ6 | |
Synthetic 15 amino acid peptide from N-terminus of human KCNJ6 / GIRK2. | |
50 μg | |
Primary | |
Canine, Equine, Hamster, Human, Mouse, Non-human Primate, Porcine, Rat | |
Antibody | |
IgG |