Learn More
KCNN3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA541014
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: ITELNDRSED LEKQIGSLES KLEHLTASFN SLPLLIADTL RQQQQQLLSA Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%
KCNN3 Polyclonal antibody specifically detects KCNN3 in Human, Mouse, Non-human Primate samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
KCNN3 | |
Polyclonal | |
Unconjugated | |
KCNN3 | |
hSK3; K3; KCa2.3; Kcnn3; potassium calcium-activated channel subfamily N member 3; potassium channel, calcium activated intermediate/small conductance subfamily N alpha, member 3; potassium intermediate/small conductance calcium-activated channel, subfamily N, member 3; SK3; SKCa 3; SKCa3; small conductance calcium-activated potassium channel 3; small conductance calcium-activated potassium channel protein 3; small-conductance Ca2+-activated K+ channel 3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
140493, 3782 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P58391, Q9UGI6 | |
KCNN3 | |
Synthetic peptide directed towards the C-terminal of human KCNN3 | |
100 μL | |
Primary | |
Human, Mouse, Non-human Primate | |
Antibody | |
IgG |