missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNQ1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA519093
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology.
KCNQ1 Polyclonal antibody specifically detects KCNQ1 in Human, Rat samples. It is validated for Western Blot.Spezifikation
KCNQ1 | |
Polyclonal | |
Unconjugated | |
KCNQ1 | |
ATFB1; ATFB3; AW559127; FLJ26167; IKs producing slow voltage-gated potassium channel subunit alpha KvLQT1; JLNS1; KCNA8; KCNA9; Kcnq1; kidney and cardiac voltage dependend K+ channel; KQT-like 1; Kv1.9; Kv7.1; KVLQT1; LQT; LQT1; mutant potassium voltage-gated channel KQT-like subfamily member 1; potassium channel, voltage gated KQT-like subfamily Q, member 1; potassium channel, voltage-gated KQT-like subfamily Q, member 1; potassium voltage-gated channel subfamily KQT member 1; potassium voltage-gated channel subfamily Q member 1; potassium voltage-gated channel, KQT-like subfamily, member 1; potassium voltage-gated channel, subfamily Q, member 1; RWS; slow delayed rectifier channel subunit; SQT2; voltage-gated potassium channel subunit Kv7.1; WRS | |
Goat | |
Antigen affinity chromatography | |
RUO | |
3784, 84020 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P51787, Q9Z0N7 | |
KCNQ1 | |
Synthetic peptide sequence (EQLTVPRRGPDEGS) corresponding to the C-terminus amino acids of KCNQ1 | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG |