missing translation for 'onlineSavingsMsg'
Learn More

KCNS2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA529511

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 13403667

  • 514.00 € / 100 Mikroliter
Verfügbar ab: 22-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Recommended positive controls: HeLa. Predicted reactivity: Mouse (99%), Rat (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

KCNS2 Polyclonal antibody specifically detects KCNS2 in Human samples. It is validated for Western Blot.
Spezifikation

Spezifikation

KCNS2
Polyclonal
Unconjugated
KCNS2
Delayed-rectifier K(+) channel alpha subunit 2; E130006J24Rik; K+ voltage-gated channel, subfamily S, 2; KCNS2; KIAA1144; Kv9.2; potassium channel, alpha subunit (kv9.2 gene); potassium voltage-gated channel modifier subfamily S member 2; potassium voltage-gated channel subfamily S member 2; potassium voltage-gated channel, delayed-rectifier, subfamily S, member 2; potassium voltage-gated channel, modifier subfamily S, member 2; voltage-gated potassium channel subunit Kv9.2
Rabbit
Antigen affinity chromatography
RUO
3788
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.66 mg/mL
PBS with 20% glycerol and 0.01% thimerosal; pH 7
Q9ULS6
KCNS2
Recombinant fragment corresponding to a region within amino acids 175 and 470 of Human KCNS2
100 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote