missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCTD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA524877
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with bovine, mouse and rat based on sequence homology.
KCTD1 Polyclonal antibody specifically detects KCTD1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
KCTD1 | |
Polyclonal | |
Unconjugated | |
KCTD1 | |
BTB/POZ domain-containing protein KCTD1; C18orf5; KCTD1; potassium channel tetramerisation domain containing 1; potassium channel tetramerization domain containing 1; potassium channel tetramerization domain-containing protein 1; SENS | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
284252 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q719H9 | |
KCTD1 | |
KLH conjugated synthetic peptide between 216-244 amino acids from the C-terminal region of human KCTD1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |