missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCTD11 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA541333
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: ADFYQIRPLL DALRELEASQ GTPAPTAALL HADVDVSPRL VHFSARRGPH Sequence homology: Human: 100%; Mouse: 90%
KCTD11 Polyclonal antibody specifically detects KCTD11 in Human samples. It is validated for Western Blot.Spezifikation
KCTD11 | |
Polyclonal | |
Unconjugated | |
KCTD11 | |
AF465352; BTB/POZ domain-containing protein KCTD11; C17orf36; KCASH1; KCASH1 protein; KCTD11; potassium channel tetramerisation domain containing 11; potassium channel tetramerization domain containing 11; potassium channel tetramerization domain-containing protein 11; REN; REN/KCTD11; retinoic acid, EGF, and NGF upregulated; retinoic acid, EGF, NGF induced gene protein; retinoic acid, EGF, NGF induced gene/potassium channel tetramerization domain containing 11; RING-type E3 ubiquitin transferase KCTD11; RING-type E3 ubiquitin transferase subunit KCTD11 | |
Rabbit | |
Protein A | |
RUO | |
147040 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q693B1 | |
KCTD11 | |
Synthetic peptide directed towards the N-terminal of human KCTD11 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |