Learn More
KCTD3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA531668
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: PC-3. Predicted reactivity: Mouse (82%), Rhesus Monkey (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
KCTD3 Polyclonal antibody specifically detects KCTD3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
KCTD3 | |
Polyclonal | |
Unconjugated | |
KCTD3 | |
4930438A20Rik; 9330185B06; BTB/POZ domain-containing protein KCTD3; E330032J19Rik; Kctd3; LOW QUALITY PROTEIN: BTB/POZ domain-containing protein KCTD3; NY-REN-45; NY-REN-45 antigen; potassium channel tetramerisation domain containing 3; potassium channel tetramerization domain containing 3; Ren-45; renal carcinoma antigen NY-REN-45; si:dkey-46n18.2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51133 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.77 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9Y597 | |
KCTD3 | |
Recombinant fragment corresponding to a region within amino acids 569 and 815 of Human KCTD3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |