missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGF Receptor 2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA524710
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
VEGF Receptor 2 Polyclonal antibody specifically detects VEGF Receptor 2 in Human, Mouse samples. It is validated for Flow Cytometry, Western Blot.Spezifikation
VEGF Receptor 2 | |
Polyclonal | |
Unconjugated | |
KDR | |
6130401C07; CD309; CD309 antigen; fetal liver kinase 1; fetal liver kinase-1; FLK1; Flk-1; FLK1 kinase insert domain receptor (a type III receptor tyrosine kinase) (VEGF receptor 2); FLK1 kinase insert domain receptor (VEGF receptor 2); flk-1 type VEGF receptor; KDR; kinase insert domain protein receptor; kinase insert domain receptor; kinase insert domain receptor (a type III receptor tyrosine kinase); kinase NYK; Krd-1; Ly73; protein-tyrosine kinase receptor flk-1; sCD309; soluble CD309; soluble vascular endothelial growth factor receptor 2; soluble VEGFR 2; soluble VEGFR2; sVEGFR-2; tyrosine kinase growth factor receptor; vascular endotheial growth factor 2; vascular endothelial growth factor receptor 2; vascular endothelial growth factor receptor- 2; vascular endothelial growth factor receptor-2; vascular endothelial growth factor receptor-3; VEGF R2; VEGF receptor-2; VEGFR; VEGFR2; VEGFR-2 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
16542, 3791 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
0.42 mg/mL | |
PBS with 0.09% sodium azide | |
P35918, P35968 | |
KDR | |
KLH conjugated synthetic peptide between 1153-1182 amino acids from human KDR | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |