missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KELL Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA547221
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
In direct ELISAs, approximately 25% cross-reactivity with recombinant mouse Kell is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
KELL Polyclonal antibody specifically detects KELL in Human samples. It is validated for Flow Cytometry, Immunoprecipitation, Western Blot.Spezifikation
KELL | |
Polyclonal | |
Unconjugated | |
KEL | |
CD238; ECE3; Kel; Kel blood group protein; Kell blood group; kell blood group antigen; kell blood group glycoprotein; Kell blood group glycoprotein homolog; Kell blood group, metallo-endopeptidase; Kell protein; RGD1565470 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
3792 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P23276 | |
KEL | |
Mouse myeloma cell line NS0-derived recombinant human Kell (aa 68-732) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |