missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KELL Monoclonal Antibody (BRIC 203), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA125131
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA1-25131 detects KELL from human samples.
KELL Monoclonal antibody specifically detects KELL in Human samples. It is validated for Flow Cytometry, Hemagglutination Assay, Immunoprecipitation.Spezifikation
KELL | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
P23276 | |
KEL | |
Human erythrocytes. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Hemagglutination Assay, Immunoprecipitation | |
BRIC 203 | |
Unconjugated | |
KEL | |
CD238; ECE3; Kel; Kel blood group protein; Kell blood group; kell blood group antigen; kell blood group glycoprotein; Kell blood group glycoprotein homolog; Kell blood group, metallo-endopeptidase; Kell protein; RGD1565470 | |
Mouse | |
Protein G | |
RUO | |
3792 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |