Learn More
Ki-67 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA519462
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Heat-mediated antigen retrieval is recommended prior to tissue staining. Recommended positive controls: ICC/IF - Wildtype HAP1 cells, SK-N-SH cells, HeLa cells, MEF1 cells IHC (P) - Mouse and human spleen tissue, human skin carcinoma tissue, human colon tissue, mouse tumor tissue, human skin tissue
Ki-67 Polyclonal antibody specifically detects Ki-67 in Human, Mouse, Rabbit samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
Ki-67 | |
Polyclonal | |
Unconjugated | |
Mki67 | |
antigen identified by monoclonal antibody Ki 67; antigen identified by monoclonal antibody Ki-67; Antigen identified by monoclonal antibody Ki-67 homolog; Antigen KI-67; Antigen KI-67 homolog; antigen KI-67; proliferation marker protein Ki-67; antigen KI-67-like; cb31; D630048A14Rik; I79_022666; Ki67; Ki-67; KIA; LOW QUALITY PROTEIN: proliferation marker protein Ki-67; marker of proliferation Ki-67; MIB-; MIB-1; Mki67; PPP1R105; Proliferation marker protein Ki-67; proliferation-related Ki-67 antigen; protein phosphatase 1, regulatory subunit 105; RP11-380J17.2; sb:cb31; si:ch211-250b22.7; unnamed protein product; wu:fa11g09; wu:fb57a07; wu:fi14e05 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100343428, 17345, 4288 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
1.0 mg/mL | |
PBS with 0.02% sodium azide; pH 7.4 | |
E9PVX6, P46013 | |
Mki67 | |
Synthetic peptide conjugated to KLH derived from within residues 1200 - 1300 of Human Ki67. | |
100 μg | |
Primary | |
Human, Mouse, Rabbit | |
Antibody | |
IgG |