missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIAA0040 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA541556
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: DPKVALESLY PLAKFTAELG LSPDKAAEVL SQGAHLAAGP DGRTIDRFSP Sequence homology: Human: 100%
KIAA0040 Polyclonal antibody specifically detects KIAA0040 in Human samples. It is validated for Western Blot.Spezifikation
KIAA0040 | |
Polyclonal | |
Unconjugated | |
KIAA0040 | |
k0040; KIAA0040; uncharacterized protein KIAA0040 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
9674 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q15053 | |
KIAA0040 | |
Synthetic peptide directed towards the middle region of human KIAA0040 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |