Learn More
KIAA0317 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA531351
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: U87-MG. Predicted reactivity: Human (99%), Mouse (98%), Rat (99%), Rhesus Monkey (99%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
KIAA0317 Polyclonal antibody specifically detects KIAA0317 in Human samples. It is validated for Western Blot.Spezifikation
KIAA0317 | |
Polyclonal | |
Unconjugated | |
AREL1 | |
apoptosis resistant E3 ubiquitin protein ligase 1; apoptosis-resistant E3 ubiquitin protein ligase 1; Apoptosis-resistant HECT-type E3 ubiquitin transferase 1; AREL1; fibrosis-inducing E3 ligase 1; FIEL1; KIAA0317; protein KIAA0317 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9870 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
O15033 | |
AREL1 | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 196 of Human KIAA0317 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |