missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EFCAB14 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA521424
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
EFCAB14 Polyclonal antibody specifically detects EFCAB14 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
EFCAB14 | |
Polyclonal | |
Unconjugated | |
EFCAB14 | |
4732418C07Rik; EFCAB14; EF-hand calcium binding domain 14; EF-hand calcium-binding domain-containing protein 14; Kiaa0494; RGD1310351; RP11-8J9.3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9813 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.59 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
O75071 | |
EFCAB14 | |
Recombinant fragment corresponding to a region within amino acids 243 and 495 of KIAA0494 (Uniprot ID#O75071) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |