Learn More
KIAA1530 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA528497
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (83%), Rat (83%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
KIAA1530 Polyclonal antibody specifically detects KIAA1530 in Human samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
KIAA1530 | |
Polyclonal | |
Unconjugated | |
UVSSA | |
KIAA1530; UV stimulated scaffold protein A; UVSS3; UVSSA; UV-stimulated scaffold protein A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57654 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q2YD98 | |
UVSSA | |
Recombinant fragment corresponding to a region within amino acids 1 and 192 of Human KIAA1530 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |