missing translation for 'onlineSavingsMsg'
Learn More

KIDINS220 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA14229

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11568780

  • 473.00 € / 100 Mikrogramm
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

PA1-4229 detects the Kidins220 protein in human samples. PA1-4229 has been successfully used in Western blot procedures. The PA1-4229 immunogen is a synthetic peptide corresponding to residues A(1747) S S E S T G F G E E R E S I L(1762) of the human 220 kDa Kidins220 protein.

KIDINS220 Polyclonal antibody specifically detects KIDINS220 in Human samples. It is validated for Western Blot.
Spezifikation

Spezifikation

KIDINS220
Polyclonal
Unconjugated
Kidins220
3110039L19Rik; AI194387; AI316525; ankyrin repeat-rich membrane spanning; ankyrin repeat-rich membrane spanning protein; ankyrin repeat-rich membrane-spanning protein; ARMS; C330002I19Rik; KIAA1250; KIDINS220; kinase D-interacting substance 220; kinase D-interacting substance of 220 kDa; kinase D-interacting substrate 220; kinase D-interacting substrate 220kDa; kinase D-interacting substrate of 220 kDa; kinase D-interacting substrate, 220kDa; MGC163482; mKIAA1250; truncated KIDINS220
Rabbit
Antigen affinity chromatography
RUO
57498
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Liquid
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q9ULH0
Kidins220
Synthetic peptide corresponding to residues A(1747) S S E S T G F G E E R E S I L(1762) of human Kidins220.
100 μg
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote