Learn More
KIF17 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA530778
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: U87-MG. Predicted reactivity: Human (99%), Mouse (88%), Bovine (84%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
KIF17 Polyclonal antibody specifically detects KIF17 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
KIF17 | |
Polyclonal | |
Unconjugated | |
Kif17 | |
5930435E01Rik; AW492270; KIAA1405; Kif17; KIF17 variant protein; Kif17b; KIF3-related motor protein; KIF3X; kinesin 17; kinesin family member 17; kinesin-like protein KIF17; KLP-2; mKIAA1405; MmKIF17; N-4 kinesin; OSM-3; RGD1562511 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57576 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.61 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9P2E2 | |
Kif17 | |
Recombinant fragment contain a sequence corresponding to a region within amino acids 780 and 1004 of KIF17 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |