Learn More
KIF1C Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA527657
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: A549, HepG2, HCT116. Predicted reactivity: Mouse (98%), Rat (96%), Rhesus Monkey (100%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
KIF1C Polyclonal antibody specifically detects KIF1C in Human samples. It is validated for Western Blot.Spezifikation
KIF1C | |
Polyclonal | |
Unconjugated | |
KIF1C | |
B430105J22Rik; D11Bwg1349e; KIAA0706; Kif1c; Kif1d; kinesin 1C; kinesin family member 1C; kinesin superfamily protein 1C; kinesin-like protein KIF1C; Kinesin-like protein KIF1D; lethal factor toxin susceptibility 1; Ltxs1; N-3 kinsin; SATX2; SAX2; spastic ataxia 2 (autosomal recessive); SPAX2; SPG58 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10749 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
O43896 | |
KIF1C | |
Recombinant fragment corresponding to a region within amino acids 452 and 758 of Human KIF1C | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |