Learn More
KIF2A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA540802
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: IEPSPETPPP PASSAKVNKI VKNRRTVASI KNDPPSRDNR VVGSARARPS Sequence homology: Cow: 86%; Dog: 93%; Guinea Pig: 93%; Horse: 86%; Human: 100%; Mouse: 79%; Rabbit: 93%; Rat: 93%
KIF2A Polyclonal antibody specifically detects KIF2A in Human samples. It is validated for Western Blot.Spezifikation
KIF2A | |
Polyclonal | |
Unconjugated | |
Kif2a | |
C530030B14Rik; CDCBM3; HK2; hypothetical protein; KIF2; kif2a; kif2-a; kif2a {ECO:0000250; kif2a.L; kinesin family member 2A; kinesin heavy chain family, member 2; kinesin heavy chain member 2; kinesin heavy chain member 2A; kinesin heavy chain member 2A L homeolog; Kinesin, heavy chain, 2; kinesin-2; Kinesin-like protein KIF2A; kinesin-related protein XKIF2; Kns2; M-kinesin; RCJMB04_9a7; UniProtKB:O00139}; Unknown (protein for MGC:133806); XELAEV_18008243mg | |
Rabbit | |
Affinity Chromatography | |
RUO | |
3796 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O00139 | |
Kif2a | |
Synthetic peptide directed towards the N-terminal of human KIF2A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |