Learn More
KTN1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA529523
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (84%), Pig (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
KTN1 Polyclonal antibody specifically detects KTN1 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
KTN1 | |
Polyclonal | |
Unconjugated | |
KTN1 | |
CG1; CG-1 antigen; KIAA0004; Kinectin; kinectin 1; kinectin 1 (kinesin receptor); kinesin receptor; KNT; ktn1; MU-RMS-40.19 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16709, 3895 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.34 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q61595, Q86UP2 | |
KTN1 | |
Recombinant fragment corresponding to a region within amino acids 999 and 1320 of Human Kinectin 1A | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |