missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kir2.1 (KCNJ2) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA537452
Weitere Details : Netto-Gewicht : 0.00910kg
Beschreibung
This antibody detects endogenous protein at a molecular weight of 48 kDa. Purity is >95% by SDS-PAGE.
Kir2.1 (KCNJ2) Polyclonal antibody specifically detects Kir2.1 (KCNJ2) in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Kir2.1 (KCNJ2) | |
Polyclonal | |
Unconjugated | |
Kcnj2 | |
ATFB9; cardiac inward rectifier KIR2.1; cardiac inward rectifier potassium channel; EGK_08925; HHBIRK1; HHIRK 1; HHIRK1; HIRK 1; hIRK1; Inward rectifier K; inward rectifier K(+) channel Kir2.1; inward rectifier K+ channel KIR2.1; inward rectifier potassium channel; inward rectifier potassium channel 2; inwardly rectifying potassium channel Kir2.1; IRK 1; IRK1; IRK-1; IRK2; Kcnf1; KCNJ2; KIR2.1; LQT 7; LQT7; Potassium channel, inwardly rectifying subfamily J member 2; potassium channel, inwardly rectifying subfamily J, member 2; potassium inwardly rectifying channel subfamily J member 2; potassium inwardly-rectifying channel, subfamily J, member 2; potassium voltage-gated channel subfamily J member 2; RBL-IRK1; SQT 3; SQT3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16518, 29712, 3759 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P35561, P63252, Q64273 | |
Kcnj2 | |
Synthetic peptide corresponding to amino acids 90-140 of human KIR21 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |