missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIR3DL2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA545520
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: LFILLLFFLL YRWCSNKKNA AVMDQEPAGD RTVNRQDSDE QDPQEVTYAQ Sequence homology: Human: 100%
KIR3DL2 Polyclonal antibody specifically detects KIR3DL2 in Human samples. It is validated for Western Blot.Spezifikation
KIR3DL2 | |
Polyclonal | |
Unconjugated | |
Kir3dl2 | |
3DL2; CD158 antigen-like family member K; CD158K; killer cell immunoglobulin like receptor, three Ig domains and long cytoplasmic tail 2; killer cell immunoglobulin-like receptor 3DL2; killer cell immunoglobulin-like receptor, three domains, long cytoplasmic tail, 2; killer Ig receptor; killer immunoglobulin-like receptor-like 2; killer-cell immunoglobulin-like receptor; killer-cell immunoglobulin-like receptor 3DL2; KIR antigen 3DL2; KIR3DL2; KIR-3DL2; Kirl2; KIR-like receptor 2; LOC727787; MHC class I NK cell receptor; natural killer cell receptor; natural killer-associated transcript 4; NKAT4; NKAT-4; NKAT4B; p140; p70 killer cell inhibitory receptor; p70 natural killer cell receptor clone CL-5; p70 NK receptor CL-5 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
3812 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P43630 | |
Kir3dl2 | |
Synthetic peptide directed towards the C-terminal of human KIR3DL2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |