missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KISS1R Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA533878
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Percent identity with other species by BLAST analysis: Human, Gorilla, Bovine, Goat, Pig (100%) Monkey, Mouse, Bat, Dog (94%) Gibbon, Marmoset, Rat, Hamster (89%).
KISS1R Polyclonal antibody specifically detects KISS1R in Bovine, Goat, Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
KISS1R | |
Polyclonal | |
Unconjugated | |
KISS1R | |
AXOR12; CPPB1; G protein-coupled receptor 54; Gpr54; G-protein coupled receptor 54; G-protein coupled receptor OT7T175; HH8; HOT7T175; Hypogonadotropin-1; kiSS-; KiSS-1; KISS1 receptor; kiSS-1 receptor; KISS1R; KiSS-1R; Kisspeptins receptor; metastin receptor; mOT7T175; orphan G protein-coupled receptor 54; orphan G protein-coupled receptor GPR54; rOT7T175 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
106502331, 528699, 84634 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q969F8 | |
KISS1R | |
Synthetic 18 amino acid peptide from 3rd cytoplasmic domain of human KISS1R / GPR54. | |
50 μg | |
Primary | |
Bovine, Goat, Human | |
Antibody | |
IgG |