missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIZUNA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA530765
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: HCT116. Predicted reactivity: Chimpanzee (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
KIZUNA Polyclonal antibody specifically detects KIZUNA in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
KIZUNA | |
Polyclonal | |
Unconjugated | |
KIZ | |
C20orf19; Centrosomal protein kizuna; Gm114; HT013; KIZ; Kizuna; kizuna centrosomal protein; NCRNA00153; PLK1S1; polo-like kinase 1 substrate 1; RGD1307071; RP69 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55857 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.46 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q2M2Z5 | |
KIZ | |
Recombinant fragment corresponding to a region within amino acids 187 and 438 of Human KIZUNA | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |