Learn More
KLB Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA544023
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: DAYTIRRGLF YVDFNSKQKE RKPKSSAHYY KQIIRENGFS LKESTPDVQG Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 85%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 93%; Rabbit: 86%; Rat: 100%
KLB Polyclonal antibody specifically detects KLB in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
KLB | |
Polyclonal | |
Unconjugated | |
KLB | |
AV071179; Betakl; betaKlotho; beta-klotho; BKL; Klb; Klb-ps1; klotho beta; klotho beta like; klotho beta, pseudogene 1; klotho beta-like protein; MGC142213; RGD1308227 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
152831 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q86Z14 | |
KLB | |
Synthetic peptide directed towards the middle region of human KLB | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |